N4-Benzoyl-2'-deoxycytidine ≥98%

Proveedor: Thermo Fisher Scientific

H52298.03 H52298.06
H52298.03EA 113 EUR
H52298.03 H52298.06
N4-Benzoyl-2'-deoxycytidine ≥98%
N4-Benzoyl-2'-deoxycytidine

Purity: 98+% CAS No.: 4836-13-9 Molecular Formula: C16H17N3O5 Formula Weight: 331.33 MDL No.: MFCD00010115

Formula: C₁₆H₁₇N₃O₅
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010115
Núm. CAS: 4836-13-9
EINECS: 225-421-9

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR