2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 97%
Proveedor: Thermo Scientific
|
Atención
|
H53398.03
H53398.06
H53398.03EA
123
EUR
H53398.03
H53398.06
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 97%
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride
Purity: 97%
CAS No.: 1171331-39-7
Molecular Formula: C4H7N2OF3.HCl
Formula Weight: 192.57
MDL No.: MFCD09944516
Formula:
C₄H₇N₂OF₃·HCl MW: 192,57 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09944516 Núm. CAS: 1171331-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...