DSG (Disuccinimidyl glutarate) 97%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
DSG
H58208.MC
H58208.ME
H58208.MCEA
188
EUR
H58208.MC
H58208.ME
DSG (Disuccinimidyl glutarate) 97%
DSG (Disuccinimidyl glutarate)
Purity: 97%
CAS No.: 79642-50-5
Molecular Formula: C13H14N2O8
Formula Weight: 326.26
Storage Temperature: -30°C to -10°C
Beilstein Reference: 8575145
MDL No.: MFCD00153597
Formula:
C₁₃H₁₄N₂O₈ MW: 326,26 g/mol |
Número MDL:
MFCD00153597 Núm. CAS: 79642-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...