N-Boc-L-threonine methyl ester 95%
Proveedor: Thermo Fisher Scientific
H61545.06
H61545.14
H61545.06EA
86.2
EUR
H61545.06
H61545.14
N-Boc-L-threonine methyl ester 95%
N-Boc-L-threonine methyl ester
Purity: 95%
CAS No.: 79479-07-5
Molecular Formula: C10H19NO5
Formula Weight: 233.26
Physical Form: Viscous liquid
Refractive Index: 1.4500
Appearance: Yellow
Boiling Point: 206°
MDL No.: MFCD00270479
Formula:
C₁₀H₁₉NO₅ Pto. Ebullición: 206 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00270479 Núm. CAS: 79479-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...