3-Fluoro-4-cyanopyridine 95%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H61594.03
H61594.06
H61594.03EA
151
EUR
H61594.03
H61594.06
3-Fluoro-4-cyanopyridine 95%
3-Fluoro-4-cyanopyridine
Purity: 95%
CAS No.: 113770-88-0
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Appearance: White to pale yellow
UN No.: UN3439
MDL No.: MFCD04112524
Formula:
C₆H₃FN₂ MW: 122.10 g/mol |
Número MDL:
MFCD04112524 Núm. CAS: 113770-88-0 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...