N,1-Di-Boc-L-histidine dicyclohexylammonium salt 98%
Proveedor: Thermo Fisher Scientific
Sinónimos:
Boc-His(Boc)-OH.DCHA, N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, Boc-His(Boc)-OH·DCHA
H62570.06
H62570.14
H62570.06EA
50.2
EUR
H62570.06
H62570.14
N,1-Di-Boc-L-histidine dicyclohexylammonium salt 98%
N,1-Di-Boc-L-histidine dicyclohexylammonium salt
Purity: 98%
CAS No.: 31687-58-8
Molecular Formula: C16H25N3O6.C12H23N
Formula Weight: 536.70
Physical Form: Solid
Melting Point: 160-165° dec
MDL No.: MFCD06804675
Formula:
C₁₆H₂₅N₃O₆·C₁₂H₂₃N MW: 536.714 g/mol Pto. de fusión: 160…165 °C (desc.) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06804675 Núm. CAS: 31687-58-8 EINECS: 250-764-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...