H-D-Lys(Boc)-OH 98%
Proveedor: Thermo Scientific
Sinónimos:
(R)-2-Amino-6-((tert-butoxycarbonyl)amino)hexanoic acid, Nepsilon-Boc-D-lysine
H62736.MD
H62736.03
H62736.MDEA
59.5
EUR
H62736.MD
H62736.03
H-D-Lys(Boc)-OH 98%
H-D-Lys(Boc)-OH
Purity: 98%
CAS No.: 31202-69-4
Molecular Formula: C11H22N2O4
Formula Weight: 246.30
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00076958
Formula:
C₁₁H₂₂N₂O₄ MW: 246,307 g/mol |
Número MDL:
MFCD00076958 Núm. CAS: 31202-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...