2-Cyanothiazole 95%
Proveedor: Thermo Scientific
|
Atención
|
H63425.03
H63425.06
H63425.03EA
156
EUR
H63425.03
H63425.06
2-Cyanothiazole 95%
2-Cyanothiazole
Purity: 95%
CAS No.: 1452-16-0
Molecular Formula: C4H2N2S
Formula Weight: 110.14
Physical Form: Solid
UN No.: UN3439
MDL No.: MFCD09702472
Formula:
C₄H₂N₂S MW: 110,14 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09702472 Núm. CAS: 1452-16-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...