3-Nitro-D-phenylalanine 95%
Proveedor: Thermo Scientific
Sinónimos:
H-D-Phe(3-NO₂)-OH, (R)-2-Amino-3-(3-nitrophenyl)propanoic acid
H63483.MD
H63483.03
H63483.MDEA
79.9
EUR
H63483.MD
H63483.03
3-Nitro-D-phenylalanine 95%
3-Nitro-D-phenylalanine
Purity: 95%
CAS No.: 169530-97-6
Molecular Formula: C9H10N2O4
Formula Weight: 210.19
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Off-white to pale yellow
MDL No.: MFCD04117829
Formula:
C₉H₁₀N₂O₄ MW: 210,189 g/mol |
Número MDL:
MFCD04117829 Núm. CAS: 169530-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...