H-D-Asp-OMe 98%
Proveedor: Thermo Fisher Scientific
Sinónimos:
(R)-3-Amino-4-methoxy-4-oxobutanoic acid, D-Aspartic acid 1-methyl ester
H63658.06
H63658.14
H63658.06EA
108
EUR
H63658.06
H63658.14
H-D-Asp-OMe 98%
H-D-Asp-OMe
Purity: 98%
CAS No.: 65414-78-0
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD03424227
Formula:
C₅H₉NO₄ MW: 147.13 g/mol |
Número MDL:
MFCD03424227 Núm. CAS: 65414-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...