3-cyclopropyl-1H-pyrazole 98%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
3-Cyclopropyl-1H-pyrazole
H64069.03
H64069.06
H64069.03EA
97.6
EUR
H64069.03
H64069.06
3-cyclopropyl-1H-pyrazole 98%
3-cyclopropyl-1H-pyrazole
Purity: 98%
CAS No.: 100114-57-6
Molecular Formula: C6H8N2
Formula Weight: 108.14
Physical Form: Liquid
Appearance: Colourless
Beilstein Reference: 1561659
MDL No.: MFCD04970938
Formula:
C₆H₈N₂ MW: 108,144 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD0497093 Núm. CAS: 100114-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...