2-Chloro-4-Bromo-5-fluorobenzaldehyde 98%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H64238.03
H64238.06
H64238.03EA
58
EUR
H64238.03
H64238.06
2-Chloro-4-Bromo-5-fluorobenzaldehyde 98%
2-Chloro-4-Bromo-5-fluorobenzaldehyde
Purity: 98%
CAS No.: 1214386-29-4
Molecular Formula: C7H3BrClFO
Formula Weight: 237.45
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD13185877
Formula:
C₇H₃BrClFO |
Número MDL:
MFCD13185877 Núm. CAS: 1214386-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...