4-methoxy-1-naphthoic acid 98%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
4-Methoxynaphthalene-1-carboxylic acid, 4-Methoxy-1-naphthoic acid
H64432.03
H64432.06
H64432.14
H64432.03EA
61.4
EUR
H64432.03
H64432.06
H64432.14
4-methoxy-1-naphthoic acid 98%
4-methoxy-1-naphthoic acid
Purity: 98%
CAS No.: 13041-62-8
Molecular Formula: C12H10O3
Formula Weight: 202.21
Physical Form: Crystalline solid
Appearance: Off-white
Melting Point: 243-247°
Beilstein Reference: 2099800
MDL No.: MFCD02575372
Formula:
C₁₂H₁₀O₃ MW: 202,209 g/mol Pto. de fusión: 243…247 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02575372 Núm. CAS: 13041-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...