3-Amino-2-pyridone 98%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone
H64480.03
H64480.06
H64480.03EA
88.4
EUR
H64480.03
H64480.06
3-Amino-2-pyridone 98%
3-Amino-2-pyridone
Purity: 98%
CAS No.: 33630-99-8
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline solid
Appearance: Brown
Beilstein Reference: 109849
MDL No.: MFCD03411556
Formula:
C₅H₆N₂O MW: 110,12 g/mol |
Número MDL:
MFCD03411556 Núm. CAS: 33630-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...