1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
7-Deazahypoxanthine
H64537.03
H64537.06
H64537.14
H64537.03EA
40.2
EUR
H64537.03
H64537.06
H64537.14
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one
Purity: 97%
CAS No.: 3680-71-5
Molecular Formula: C6H5N3O
Formula Weight: 135.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture and Light Sensitive
Appearance: Brown
Melting Point: 345-350°
Beilstein Reference: 607869
MDL No.: MFCD08234923
Formula:
C₆H₅N₃O MW: 135,13 g/mol Pto. de fusión: 345…350 °C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD08234923 Núm. CAS: 3680-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...