2-Amino-6-cloro-3-nitropiridina 98%
Proveedor: Thermo Scientific
|
Peligro
|
Sinónimos:
6-Cloro-3-nitro-2-piridilamina
H64634.14
H64634.22
H64634.14EA
91.7
EUR
H64634.14
H64634.22
2-Amino-6-cloro-3-nitropiridina 98%
2-Amino-6-cloro-3-nitropiridina
Purity: 98%
CAS No.: 27048-04-0
Molecular Formula: C5H4ClN3O2
Formula Weight: 173.56
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Appearance: Yellow
Melting Point: 192-196°
Beilstein Reference: 474607
UN No.: UN2811
MDL No.: MFCD00044350
Formula:
C₅H₄ClN₃O₂ MW: 173,56 g/mol Pto. de fusión: 192…196 °C |
Número MDL:
MFCD00044350 Núm. CAS: 27048-04-0 EINECS: 248-188-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...