2-Amino-5-fluoro-6-methylpyridine 98%
Proveedor: Thermo Scientific
|
Atención
|
H64700.03
H64700.06
H64700.03EA
63.4
EUR
H64700.03
H64700.06
2-Amino-5-fluoro-6-methylpyridine 98%
2-Amino-5-fluoro-6-methylpyridine
Purity: 98%
CAS No.: 110919-71-6
Molecular Formula: C6H7FN2
Formula Weight: 126.13
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 4376910
UN No.: UN2811
MDL No.: MFCD09835298
Formula:
C₆H₇FN₂ MW: 126,13 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09835298 Núm. CAS: 110919-71-6 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...