4-(9-Carbazolyl)benzeneboronic acid 98%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
4-(9H-9-carbozale)phenylboronic acid, 4-(9H-Carbazol-9-yl)phenylboronic acid, (4-(9H-Carbazol-9-yl)phenyl)boronic acid, 4-(9H-Carbazol-9-yl)benzeneboronic acid
H64703.03
H64703.06
H64703.03EA
107
EUR
H64703.03
H64703.06
4-(9-Carbazolyl)benzeneboronic acid 98%
4-(9-Carbazolyl)benzeneboronic acid
Purity: 98%
CAS No.: 419536-33-7
Molecular Formula: C18H14BNO2
Formula Weight: 287.13
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 10565612
MDL No.: MFCD13176534
Formula:
C₁₈H₁₄BNO₂ MW: 287,125 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD13176534 Núm. CAS: 419536-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...