3-Fluoro-2-methylpyridine 98%
Proveedor: Thermo Fisher Scientific
|
Peligro
|
H64705.03
H64705.06
H64705.03EA
120
EUR
H64705.03
H64705.06
3-Fluoro-2-methylpyridine 98%
3-Fluoro-2-methylpyridine
Purity: 98%
CAS No.: 15931-15-4
Molecular Formula: C6H6FN
Formula Weight: 111.12
Physical Form: Liquid
Appearance: Colourless to pale yellow
Beilstein Reference: 6497832
MDL No.: MFCD08705710
Formula:
C₆H₆FN MW: 111.119 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08705710 Núm. CAS: 15931-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...