3-fluoro-5-iodopyridine 95%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H64770.03
H64770.06
H64770.03EA
144
EUR
H64770.03
H64770.06
3-fluoro-5-iodopyridine 95%
3-Fluoro-5-iodopyridine
Purity: 95%
CAS No.: 757950-13-3
Molecular Formula: C5H3FIN
Formula Weight: 222.99
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
Beilstein Reference: 11107918
MDL No.: MFCD08457780
Formula:
C₅H₃FIN MW: 222.989 g/mol |
Número MDL:
MFCD08457780 Núm. CAS: 757950-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...