3-Oxocyclobutanecarboxylic acid 98%
Proveedor: Thermo Scientific
|
Atención
|
H64774.06
H64774.14
H64774.06EA
83.2
EUR
H64774.06
H64774.14
3-Oxocyclobutanecarboxylic acid 98%
3-Oxocyclobutanecarboxylic acid
Purity: 98%
CAS No.: 23761-23-1
Molecular Formula: C5H6O3
Formula Weight: 114.10
Physical Form: Solid
Appearance: Off-white
Melting Point: 69-70°
Beilstein Reference: 2078401
MDL No.: MFCD00100900
Formula:
C5H6O3 MW: 114,10 g/mol Pto. de fusión: 69…70 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00100900 Núm. CAS: 23761-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...