9-Bromo-1-nonene 97%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H64792.03
H64792.06
H64792.03EA
48.2
EUR
H64792.03
H64792.06
9-Bromo-1-nonene 97%
9-Bromo-1-nonene
Purity: 97%
CAS No.: 89359-54-6
Molecular Formula: C9H17Br
Formula Weight: 205.14
Physical Form: Liquid
Refractive Index: 1.4675
Appearance: Colourless to pale yellow
Boiling Point: 100-101°/15mm
Beilstein Reference: 1746338
MDL No.: MFCD09037826
Formula:
C₉H₁₇Br MW: 205.139 g/mol Pto. Ebullición: 100…101 °C (15 mmHg) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09037826 Núm. CAS: 89359-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...