Methyl-4-methylsalicylate 98+%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
Methyl 2-hydroxy-4-methylbenzoate
H64793.06
H64793.14
H64793.06EA
55.3
EUR
H64793.06
H64793.14
Methyl-4-methylsalicylate 98+%
Methyl-4-methylsalicylate
Purity: 98+%
CAS No.: 4670-56-8
Molecular Formula: C9H10O3
Formula Weight: 166.18
Storage Temperature: 2°C to 8°C
Physical Form: Low melting solid
Density: 1.147
Refractive Index: 1.5370
Appearance: Pale brown to pale yellow
Melting Point: 27-28°
Boiling Point: 242° lit.
Beilstein Reference: 2690247
MDL No.: MFCD00020130
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol Pto. Ebullición: 242 °C (lit.) Pto. de fusión: 27…28 °C Densidad: 1,147 |
Número MDL:
MFCD00020130 Núm. CAS: 4670-56-8 EINECS: 225-117-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...