3-Fluoro-2-formylphenylboronic acid 95%
Proveedor: Thermo Scientific
|
Peligro
|
Sinónimos:
(3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid
Purity: 95%
CAS No.: 871126-15-7
Molecular Formula: C7H6BFO3
Formula Weight: 167.93
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 123-127°
UN No.: UN3261
MDL No.: MFCD10697421
Formula:
C₇H₆BFO₃ MW: 167,93 g/mol Pto. de fusión: 123…127 °C |
Número MDL:
MFCD10697421 Núm. CAS: 871126-15-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...