Fmoc-D-allo-thr(tBu)-OH 98%
Proveedor: Thermo Scientific
Sinónimos:
N-Fmoc-O-tert-butyl-D-allo-threonine, Fmoc-D-Allo-Thr(tBu)-OH
H66486.03
H66486.03EA
237
EUR
H66486.03
Fmoc-D-allo-thr(tBu)-OH 98%
Fmoc-D-allo-thr(tBu)-OH
Purity: 98%
CAS No.: 170643-02-4
Molecular Formula: C23H27NO5
Formula Weight: 397.47
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00077074
Formula:
C₂₃H₂₇NO₅ MW: 397,471 g/mol |
Número MDL:
MFCD00077074 Núm. CAS: 170643-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...