β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%

Proveedor: Thermo Scientific

Sinónimos: β-NADPH tetrasodium salt tetrahydrate

J60387.MC J60387.ME
J60387.MCEA 243 EUR
J60387.MC J60387.ME
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate

Applications: beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.


  • Store in cool place
  • Keep container tightly closed in a dry and well-ventilated place
  • Light sensitive
  • Hygroscopic


Purity: 98+% Molecular Formula: C21H26N7Na4O17P3.xH2O Formula Weight: 905.42 (833.35anhy) Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD07366294

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR