Carboplatin
Proveedor: Thermo Scientific
|
Peligro
|
Sinónimos:
cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)
J60433.03
J60433.06
J60433.03EA
425
EUR
J60433.03
J60433.06
Carboplatin
Carboplatin
CAS No.: 41575-94-4
Molecular Formula: C6H12N2O4Pt
Formula Weight: 371.25
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 10mg/ml
MDL No.: MFCD00070464
Applications: An antitumor agent
Formula:
C₆H₁₂N₂O₄Pt MW: 371,26 g/mol |
Número MDL:
MFCD00070464 Núm. CAS: 41575-94-4 EINECS: 255-446-0 Índice Merck: 14,1822 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...