Hexamethonium bromide ≥98%
Proveedor: Thermo Fisher Scientific
Atención
|
Purity: 98+%
CAS No.: 55-97-0
Molecular Formula: C12H30Br2N2
Formula Weight: 362.19
Physical Form: Powder
Appearance: White
Melting Point: 282-285° dec.
Beilstein Reference: 3715749
MDL No.: MFCD00011787
Applications: Nicotinic acetyl choline receptor antagonist
Formula:
C₁₂H₃₀Br₂N₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011787 Núm. CAS: 55-97-0 EINECS: 200-249-7 Índice Merck: 14,4688 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...