Metoclopramide hydrochloride
Proveedor: Thermo Scientific
|
Atención
|
J61545.09
J61545.14
J61545.22
J61545.09EA
48
EUR
J61545.09
J61545.14
J61545.22
Metoclopramide hydrochloride
Metoclopramide hydrochloride
CAS No.: 54143-57-6
Molecular Formula: C14H22ClN3O2.HCl
Formula Weight: 336.26
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline solid
Sensitivity: Light Sensitive
Appearance: White to pale yellow
MDL No.: MFCD00150634
Applications: 5-HT3 serotonin antagonist; D2 dopamine antagonist
Formula:
C₁₄H₂₂ClN₃O₂·HCl MW: 336,26 g/mol |
Número MDL:
MFCD00058011 Núm. CAS: 7232-21-5 Índice Merck: 14,6143 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...