DL-Aminoglutethimide 99%

Proveedor: Thermo Scientific
Atención

Sinónimos: 3-(4-Aminophenyl)-3-ethylpiperidine-2,6-dione, Aminoglutethimide, 3-(p-Aminophenyl)-3-ethylpiperidine-2,6-dione,3-(4-Aminophenyl)-3-ethyl-2,6-piperidinedione

J62182.03 J62182.06 J62182.14
J62182.03EA 35.6 EUR
J62182.03 J62182.06 J62182.14
DL-Aminoglutethimide 99%
DL-Aminoglutethimide

Purity: 99% CAS No.: 125-84-8 Molecular Formula: C13H16N2O2 Formula Weight: 232.28 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 152-154° MDL No.: MFCD00010122


Applications: Adrenocortical suppressant which also inhibits conversion of androgens to estrogens by the aromatase enzyme system

Formula: C₁₃H₁₆N₂O₂
MW: 232,28 g/mol
Pto. de fusión: 152…154 °C
Número MDL: MFCD00010122
Núm. CAS: 125-84-8
EINECS: 204-756-4
Índice Merck: 14,440

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR