Norfloxacin
Proveedor: Thermo Fisher Scientific
Sinónimos:
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
J62652.03
J62652.06
J62652.14
J62652.03EA
33.7
EUR
J62652.03
J62652.06
J62652.14
Norfloxacin
Norfloxacin
CAS No.: 70458-96-7
Molecular Formula: C16H18FN3O3
Formula Weight: 319.33
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 220-221°
Solubility: Soluble in acetic acid. Also soluble in acetone or cloroform. Slightly soluble in water
MDL No.: MFCD00079532
Applications: A synthetic fluoroquinolone antibiotic
Formula:
C₁₆H₁₈FN₃O₃ Pto. de fusión: 220…221 °C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00079532 Núm. CAS: 70458-96-7 EINECS: 274-614-4 Índice Merck: 14,6700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...