Zeatin riboside
Proveedor: Thermo Scientific
Sinónimos:
trans-Zeatinriboside, trans-Zeatin-riboside
J63382.MB
J63382.MF
J63382.MC
J63382.MBEA
199
EUR
J63382.MB
J63382.MF
J63382.MC
Zeatin riboside
Zeatin riboside
CAS No.: 6025-53-2
Molecular Formula: C15H21N5O5
Formula Weight: 351.36
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 5460894
MDL No.: MFCD00036809
Applications: Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence
Formula:
C₁₅H₂₁N₅O₅ MW: 351,363 g/mol |
Número MDL:
MFCD00036809 Núm. CAS: 6025-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...