Imipramine hydrochloride

Proveedor: Thermo Scientific
Atención

Sinónimos: 3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine hydrochloride

J63723.06 J63723.14
J63723.06EA 70.2 EUR
J63723.06 J63723.14
Imipramine hydrochloride
Imipramine hydrochloride

CAS No.: 113-52-0 Molecular Formula: C19H24N2.HCl Formula Weight: 316.87 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White Melting Point: 174-175° Solubility: Soluble in water MDL No.: MFCD00012669


Applications: A serotonin uptake inhibitor

Formula: C₁₉H₂₄N₂·HCl
MW: 316,87 g/mol
Pto. de fusión: 174…175 °C
Número MDL: MFCD00012669
Núm. CAS: 113-52-0
EINECS: 204-030-7
Índice Merck: 14,4920

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR