Difloxacin hydrochloride
Proveedor: Thermo Scientific
Sinónimos:
6-Fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
J63754.06
J63754.14
J63754.06EA
139
EUR
J63754.06
J63754.14
Difloxacin hydrochloride
Difloxacin hydrochloride
CAS No.: 91296-86-5
Molecular Formula: C21H19F2N3O3.HCl
Formula Weight: 435.85
Physical Form: Powder
Appearance: White
Solubility: Soluble in aqueous buffer
Beilstein Reference: 6464798
MDL No.: MFCD03840489
Applications: A quinolone antibacterial compound. Structurally related to norfloxacin
Formula:
C₂₁H₁₉F₂N₃O₃·HCl MW: 435,85 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03840489 Núm. CAS: 91296-86-5 Índice Merck: 14,3141 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...