Acebutolol hydrochloride
Proveedor: Thermo Fisher Scientific
Sinónimos:
N-(3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl)butanamide, Acebutolol HCl, Acebutolol hydrochloride
J63790.03
J63790.06
J63790.09
J63790.03EA
66.8
EUR
J63790.03
J63790.06
J63790.09
Acebutolol hydrochloride
Acebutolol hydrochloride
CAS No.: 34381-68-5
Molecular Formula: C18H28N2O4.HCl
Formula Weight: 372.89
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 141-143°
MDL No.: MFCD00078860
Applications: Cardioselective beta-adrenergic blocker
Formula:
C₁₈H₂₈N₂O₄·HCl MW: 372.89 g/mol Pto. de fusión: 141…143 °C |
Número MDL:
MFCD00078860 Núm. CAS: 34381-68-5 EINECS: 251-980-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...