Dizocilpine maleate ≥99%

Proveedor: Thermo Scientific

Sinónimos: (5S,10R)-5-Methyl-10,11-dihydro-5H-5,10-epiminodibenzo[a,d][7]annulene maleate, 10,11-dihydro-5S-methyl-5H-dibenzo[a,d]cyclohepten-5,10-imine, (2Z)-2-butenedioate,Dizocilpine, (+)-MK 801 hydrogen maleate

J63917.MA J63917.MF
J63917.MAEA 121 EUR
J63917.MA J63917.MF
Dizocilpine maleate ≥99%
Dizocilpine maleate

Purity: 99+% CAS No.: 77086-22-7 Molecular Formula: C16H15N.C4H4O4 Formula Weight: 337.37 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00082465


Applications: NMDA receptor antagonist that prevents calcium flux

Formula: C₁₆H₁₅N·C₄H₄O₄
MW: 337,37 g/mol
Número MDL: MFCD00082465
Núm. CAS: 77086-22-7
EINECS: 278-614-5
Índice Merck: 14,3386

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR