1-β-D-Arabinofuranosyluracil 99%
Proveedor: Thermo Scientific
Sinónimos:
Ara-U, 1-((2R,3S,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, Uracil 1-β-D-Arabinofuranoside
J64147.06
J64147.14
J64147.06EA
47.2
EUR
J64147.06
J64147.14
1-β-D-Arabinofuranosyluracil 99%
1-beta-D-Arabinofuranosyluracil
Purity: 99%
CAS No.: 3083-77-0
Molecular Formula: C9H12N2O6
Formula Weight: 244.20
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 28749
MDL No.: MFCD00065998
Formula:
C₉H₁₂N₂O₆ MW: 244,20 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00065998 Núm. CAS: 3083-77-0 EINECS: 221-386-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...