3-Metiladenina

Proveedor: Thermo Fisher Scientific

Sinónimos: 3-Metil-3H-adenina

J64813.MB J64813.MF
J64813.MBEA 74.3 EUR
J64813.MB J64813.MF
3-Metiladenina
3-Metiladenina

A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation.


  • Autophagy Inhibitor

Formula: C₆H₇N₅
MW: 149.16 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00010531
Núm. CAS: 5142-23-4
EINECS: 225-908-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR