2-Deoxy-D-galactose 99%
Proveedor: Thermo Scientific
J65482.04
J65482.06
J65482.09
J65482.04EA
123
EUR
J65482.04
J65482.06
J65482.09
2-Deoxy-D-galactose 99%
2-Deoxy-D-galactose
Purity: 99%
CAS No.: 1949-89-9
Molecular Formula: C6H12O5
Formula Weight: 164.16
Physical Form: Powder
Appearance: White to off-white
Melting Point: 107-110°
Beilstein Reference: 1723333
MDL No.: MFCD00014649
Applications: For studying galactose uptake into Escherichia coli
Formula:
C₆H₁₂O₅ MW: 164,16 g/mol Pto. de fusión: 107…110 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00014649 Núm. CAS: 1949-89-9 EINECS: 217-765-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...