Nisin

Proveedor: Thermo Fisher Scientific

J66370.03 J66370.06 J66370.14
J66370.03EA 79.4 EUR
J66370.03 J66370.06 J66370.14
Nisin
Nisin

Effective against a broad range of Gram-positive bacteria but not against Gram-negative cell membranes


  • 900 I.U./mg


CAS No.: 1414-45-5 Molecular Formula: C143H230N42O37S7 Formula Weight: 3354.06 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: Pale brown Boiling Point: 2967° Solubility: Soluble in water MDL No.: MFCD00131724


Applications: Effective against a broad range of gram positive bacteria but not against gram negative cell membranes

Formula: C₁₄₃H₂₃₀N₄₂O₃₇S₇
Pto. Ebullición: 2967 °C
Pto. de inflamación: >110 °C (230 °F)
Número MDL: MFCD00131724
Núm. CAS: 1414-45-5
EINECS: 215-807-5
Índice Merck: 14,6564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR