2-Chloroacrylic acid ≥96%
Proveedor: Thermo Fisher Scientific
|
Peligro
|
L00350.04
L00350.09
L00350.04EA
82.8
EUR
L00350.04
L00350.09
2-Chloroacrylic acid ≥96%
2-Chloroacrylic acid
Purity: 98+%
CAS No.: 598-79-8
Molecular Formula: C3H3ClO2
Formula Weight: 106.51
Storage Temperature: -30°C to -10°C
Melting Point: 63-67°
Beilstein Reference: 1098503
UN No.: UN3261
MDL No.: MFCD00014336
Formula:
C₃H₃ClO₂ Pto. de fusión: 63…67 °C Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00014336 Núm. CAS: 598-79-8 EINECS: 209-953-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...