Polygalacturonic acid, M.W. 25,000-50,000
Proveedor: Thermo Fisher Scientific
L04206.36
L04206.22
L04206.14
L04206.36EA
903
EUR
L04206.36
L04206.22
L04206.14
Polygalacturonic acid, M.W. 25,000-50,000
Polygalacturonic acid
CAS No.: 25990-10-7
Optical Rotation: +250° (c=1 in 0.1N NaOH)
MDL No.: MFCD00131972
Temperatura de almacenaje:
Ambiente |
Número MDL:
MFCD00131972 Núm. CAS: 25990-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...