2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde ≥98%
Proveedor: Thermo Scientific
Sinónimos:
2,5-Dimethyl-1-phenyl-1H-pyrrole-3-carbaldehyde
L05836.06
L05836.14
L05836.06EA
98.7
EUR
L05836.06
L05836.14
2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde ≥98%
2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde
Purity: 98+%
CAS No.: 83-18-1
Molecular Formula: C13H13NO
Formula Weight: 199.25
Storage Temperature: 2°C to 8°C
Melting Point: 87-90°
Beilstein Reference: 144790
MDL No.: MFCD00051494
Formula:
C₁₃H₁₃NO MW: 199,25 g/mol Pto. de fusión: 87…90 °C |
Número MDL:
MFCD00051494 Núm. CAS: 83-18-1 EINECS: 201-458-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...