2-(Aminomethyl)benzimidazole dihydrochloride hydrate 98%
Proveedor: Thermo Scientific
L07440.06
L07440.14
L07440.06EA
47.7
EUR
L07440.06
L07440.14
2-(Aminomethyl)benzimidazole dihydrochloride hydrate 98%
2-(Aminomethyl)benzimidazole dihydrochloride hydrate
Purity: 98%
CAS No.: 5993-91-9
Molecular Formula: C8H9N3.2HCl.xH2O
Formula Weight: 220.10(anhy)
Melting Point: ca 264° dec.
Beilstein Reference: 3698942
MDL No.: MFCD00150073
Formula:
C₈H₉N₃·2HCl·xH₂O MW: 220,10 g/mol (anhy) Pto. de fusión: ∼ 264 °C (desc.) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00150073 Núm. CAS: 5993-91-9 EINECS: 227-820-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...