(R)-(-)-S-Methyl-S-phenylsulphoximine 97%
Proveedor: Thermo Fisher Scientific
L08562.03
L08562.MD
L08562.03EA
216
EUR
L08562.03
L08562.MD
(R)-(-)-S-Methyl-S-phenylsulphoximine 97%
(R)-(-)-S-Methyl-S-phenylsulphoximine
Purity: 97%
CAS No.: 60933-65-5
Molecular Formula: C7H9NOS
Formula Weight: 155.22
Melting Point: 29-31°
Beilstein Reference: 5245635
Optical Rotation: -36° (c=1 in acetone)
MDL No.: MFCD00151460
Formula:
C₇H₉NOS MW: 155.22 g/mol Pto. de fusión: 29…31 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00151460 Núm. CAS: 60933-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...