4-(Dimethylamino)-4-oxobutanoic acid 98%
Proveedor: Thermo Scientific
L08796.06
L08796.14
L08796.06EA
67.6
EUR
L08796.06
L08796.14
4-(Dimethylamino)-4-oxobutanoic acid 98%
4-(Dimethylamino)-4-oxobutanoic acid
Purity: 98%
CAS No.: 2564-95-6
Molecular Formula: C6H11NO3
Formula Weight: 145.16
Melting Point: 82-87°
Beilstein Reference: 1762788
MDL No.: MFCD00043505
Formula:
C₆H₁₁NO₃ Pto. de fusión: 82…87 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00043505 Núm. CAS: 2564-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...