Boc-sarcosine 99%
Proveedor: Thermo Fisher Scientific
Sinónimos:
Boc-Sar-OH
L13482.06
L13482.14
L13482.06EA
29.7
EUR
L13482.06
L13482.14
Boc-sarcosine 99%
Boc-sarcosine
Purity: 99%
CAS No.: 13734-36-6
Molecular Formula: C8H15NO4
Formula Weight: 189.20
Storage Temperature: 2°C to 8°C
Melting Point: 88-93°
Beilstein Reference: 2046827
MDL No.: MFCD00037795
Formula:
C₈H₁₅NO₄ MW: 189.211 g/mol Pto. de fusión: 88…93 °C |
Número MDL:
MFCD00037795 Núm. CAS: 13734-36-6 EINECS: 237-306-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...