4-Bromo-2-methylbiphenyl 98%
Proveedor: Thermo Fisher Scientific
L17875.03
L17875.06
L17875.03EA
30.8
EUR
L17875.03
L17875.06
4-Bromo-2-methylbiphenyl 98%
4-Bromo-2-methylbiphenyl
Purity: 98%
CAS No.: 5002-26-6
Molecular Formula: C13H11Br
Formula Weight: 247.14
Refractive Index: 1.6210
Boiling Point: 113-114°/0.7mm
Beilstein Reference: 1908937
MDL No.: MFCD03093069
Formula:
C₁₃H₁₁Br Pto. Ebullición: 113…114 °C (0,7 mmHg) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03093069 Núm. CAS: 5002-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...