Oleanolic acid, en polvo

Proveedor: MP Biomedicals

0215398190 0215398180
ICNA0215398190EA 507 EUR
ICNA0215398190 ICNA0215398180
Oleanolic acid, en polvo
Oleanolic acid

Soluble in chloroform (40 mg/4 ml - clear, colourless solution); insoluble in water.


Oleanolic acid is a antitumor and hepatoprotective agent.


It has therapeutic potential for neurodegenerative diseases.

Formula: C₃₀H₄₈O₃
Número MDL: MFCD00064914
Núm. CAS: 508-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR