Bethanechol chloride
Proveedor: MP Biomedicals
H2O: soluble 1.7 g/mL (stable for several days at 4 °C.); Ethanol: soluble 80 mg/mL (stable for several days at 4°C.) (Lit.)
Bethanechol Chloride is used to study beneficial effects of estrogen in a mouse model of cerebrovascular insufficiency.
Formula:
C₇H₁₇ClN₂O₂ |
Número MDL:
MFCD00055224 Núm. CAS: 590-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...